AC5H3165C43F Accela ChemBio Inc.

3,7-Bis[4-(tert-butyl)styryl]-10-(3,5-diiodo-4-methylphenyl)-2,8-diethyl-5,5-difluoro-1,9-dimethyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide

  • MDL number MFCD31811569
  • Molecular Weight : 934.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form Dark green powder
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248222&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248222.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart