AC5H3165C449 Accela ChemBio Inc.

Ethyl 3-Amino-1,4,5,6,7,8-hexahydrocyclohepta[b]pyrrole-2-carboxylate

  • MDL number MFCD31811614
  • Cas Number : 2148923-28-6
  • Molecular Weight : 222.28352

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249727&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249727.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart