AC5H3165C44C Accela ChemBio Inc.

Ethyl 2,3-Dimethyl-6-[3-(trifluoromethyl)benzyl]-6H-thieno[2,3-b]pyrrole-5-carboxylate

  • MDL number MFCD31811630
  • Molecular Weight : 381.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250514&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250514.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart