AC5H3165C44D Accela ChemBio Inc.

3-(3-Iodopropyl)pyridine Hydrochloride

  • MDL number MFCD31811631
  • Molecular Weight : 283.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form White to yellow solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250698&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250698.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart