AC5H3165C478 Accela ChemBio Inc.

1-(6-Chloro-3-indolyl)-2,2,2-trifluoroethanone

  • MDL number MFCD15524801
  • Cas Number : 676477-10-4
  • Molecular Weight : 247.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY058839&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY058839.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart