AC5H3165C47D Accela ChemBio Inc.

2-Ethyl-3-piperidinone Hydrochloride

  • MDL number MFCD08061213
  • Cas Number : 1159826-57-9
  • Molecular Weight : 163.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-ethyl-3-piperidinone hydrochloride
Inchi Code 1S/C7H13NO.ClH/c1-2-6-7(9)4-3-5-8-6;/h6,8H,2-5H2,1H3;1H
Inchi Key RKYYSPXEELHYRU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY064260&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY064260.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart