AC5H3165C491 Accela ChemBio Inc.

Glycodeoxycholic Acid

  • MDL number MFCD00067133
  • Cas Number : 360-65-6
  • Molecular Weight : 449.62332

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY120981&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY120981.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart