AC5H3165C493 Accela ChemBio Inc.

6-Aminopyridine-2-carbohydrazide

  • MDL number MFCD18205001
  • Cas Number : 5584-18-9
  • Molecular Weight : 152.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-2-pyridinecarbohydrazide
Inchi Code 1S/C6H8N4O/c7-5-3-1-2-4(9-5)6(11)10-8/h1-3H,8H2,(H2,7,9)(H,10,11)
Inchi Key KIZIYKSOOXNJRU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129373&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129373.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart