AC5H3165C497 Accela ChemBio Inc.

Ethyl 4-Chloro-6-methylpyrazolo[1,5-a]pyrazine-2-carboxylate

  • MDL number MFCD28143931
  • Cas Number : 1449598-75-7
  • Molecular Weight : 239.6583

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY213972&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY213972.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart