AC5H3165C499 Accela ChemBio Inc.

1-(4-Iodophenyl)cyclopentanecarbonitrile

  • MDL number MFCD01838824
  • Cas Number : 324532-35-6
  • Molecular Weight : 297.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226749&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226749.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart