AC5H3165C49A Accela ChemBio Inc.

1-(4-Iodophenyl)cyclopentanemethanamine

  • MDL number MFCD28466843
  • Cas Number : 1895253-52-7
  • Molecular Weight : 301.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-(4-iodophenyl)cyclopentyl)methanamine
Inchi Code 1S/C12H16IN/c13-11-5-3-10(4-6-11)12(9-14)7-1-2-8-12/h3-6H,1-2,7-9,14H2
Inchi Key DNVGWJCYAAJTRW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to tan solid, or colorless to tan liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226751&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226751.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart