AC5H3165C49E Accela ChemBio Inc.

Tetra-n-butylammonium Phenyltrifluoroborate

  • MDL number MFCD09953443
  • Cas Number : 149477-41-8
  • Molecular Weight : 387.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N,N,N-tributyl-1-butanaminium trifluoro(phenyl)borate(1-)
Inchi Code 1S/C16H36N.C6H5BF3/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;8-7(9,10)6-4-2-1-3-5-6/h5-16H2,1-4H3;1-5H/q+1;-1
Inchi Key SQRLCDKEHWULQU-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. +4?
Physical Form White powder or crystal
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY001021&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY001021.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart