AC5H3165C4A0 Accela ChemBio Inc.

1-Chloro-5-nitronaphthalene

  • MDL number MFCD01026465
  • Cas Number : 605-63-0
  • Molecular Weight : 207.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237477&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237477.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart