AC5H3165C4A2 Accela ChemBio Inc.

3-(3-Amino-4-nitrophenyl)-1-propanol

  • MDL number MFCD31716133
  • Cas Number : 849235-84-3
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-amino-4-nitrophenyl)propan-1-ol
Inchi Code 1S/C9H12N2O3/c10-8-6-7(2-1-5-12)3-4-9(8)11(13)14/h3-4,6,12H,1-2,5,10H2
Inchi Key HLUABOUHLBVAOZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246035&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246035.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart