AC5H3165C4A5 Accela ChemBio Inc.

(E)-2-[3-[[[3-(2-Pyridylethynyl)-2-cyclohexen-1-ylidene]amino]oxy]propoxy]ethyl Methanesulfonate

  • MDL number MFCD30741386
  • Cas Number : 1451015-35-2
  • Molecular Weight : 392.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-(((5-(pyridin-2-ylethynyl)cyclohexa-1,5-dien-1-yl)amino)oxy)propoxy)ethyl methanesulfonate

Description

Purity 95%
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247304&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247304.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart