AC5H3165C4A8 Accela ChemBio Inc.

4-Isocyanato-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD20483372
  • Cas Number : 143782-18-7
  • Molecular Weight : 212.1281296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247506&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247506.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart