AC5H3165C4AA Accela ChemBio Inc.

5-Amino-1-methyl-1H-pyrazole-3-ethanol

  • MDL number MFCD27942327
  • Cas Number : 1224888-33-8
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247720&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247720.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart