AC5H3165C4AB Accela ChemBio Inc.

5-Amino-7-(5-methyl-2-furyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidine

  • MDL number MFCD24642508
  • Cas Number : 442907-10-0
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247784&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247784.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart