AC5H3165C4AD Accela ChemBio Inc.

1-Boc-2,2-dimethyl-3-piperidinone

  • MDL number MFCD30619546
  • Cas Number : 1525448-33-2
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to yellow liquid, or white to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248058&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248058.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart