AC5H3165C4B1 Accela ChemBio Inc.

1-Cyclopropyl-1H-pyrazolo[3,4-d]pyrimidine-6-carboxylic Acid

  • MDL number MFCD31811564
  • Cas Number : 2230521-72-7
  • Molecular Weight : 204.18542

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248166&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248166.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart