AC5H3165C4B5 Accela ChemBio Inc.

2-Bromo-5-(difluoromethyl)pyridin-3-amine

  • MDL number MFCD25476839
  • Cas Number : 1804445-79-1
  • Molecular Weight : 223.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248270&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248270.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart