AC5H3165C4B7 Accela ChemBio Inc.

6-Hydroxy-1H-indazole-7-carbaldehyde

  • MDL number MFCD31811575
  • Cas Number : 1808943-48-7
  • Molecular Weight : 162.14544

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Pale yellow to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248310&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248310.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart