AC5H3165C4B8 Accela ChemBio Inc.

2-Bromo-alpha-methylpyridine-4-methanamine

  • MDL number MFCD18254221
  • Cas Number : 1211534-68-7
  • Molecular Weight : 201.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248318&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248318.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart