AC5H3165C4B9 Accela ChemBio Inc.

4-Cyclopropyl-2-fluorophenylboronic Acid Pinacol Ester

  • MDL number MFCD18732815
  • Cas Number : 1338718-13-0
  • Molecular Weight : 262.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248319&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248319.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart