AC5H3165C4BE Accela ChemBio Inc.

2-Benzyl-4,6-dichloro-1,3,5-triazine

  • MDL number MFCD22381680
  • Cas Number : 26650-79-3
  • Molecular Weight : 240.08868

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248578&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248578.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart