AC5H3165C4C7 Accela ChemBio Inc.

3,5-Dibromo-1-(methoxymethyl)-1H-1,2,4-triazole

  • MDL number MFCD27756632
  • Cas Number : 1785762-67-5
  • Molecular Weight : 270.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249394&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249394.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart