AC5H3165C4C8 Accela ChemBio Inc.

1-[3-Bromo-1-(methoxymethyl)-1H-1,2,4-triazol-5-yl]-4-[(2-tetrahydropyranyl)oxy]-1-butanone

  • MDL number MFCD31758455
  • Molecular Weight : 362.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249395&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249395.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart