AC5H3165C4CC Accela ChemBio Inc.

5-Bromo-6-methoxyisoindolin-1-one

  • MDL number MFCD23707902
  • Cas Number : 1427356-93-1
  • Molecular Weight : 242.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249523&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249523.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart