AC5H3165C4CD Accela ChemBio Inc.

6,10,14-Trimethylpentadecan-2-one

  • MDL number MFCD00065420
  • Cas Number : 502-69-2
  • Molecular Weight : 268.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249550&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249550.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart