AC5H3165C4CF Accela ChemBio Inc.

3-(4-Fluorobenzoyl)benzaldehyde

  • MDL number MFCD19320303
  • Molecular Weight : 228.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249573&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249573.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart