AC5H3165C4D3 Accela ChemBio Inc.

2,2-Dimethoxy-N-(4-nitrobenzyl)ethanamine

  • MDL number MFCD12567444
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249612&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249612.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart