AC5H3165C4DB Accela ChemBio Inc.

4,5-Dihydroxy-3,5-octadiene-2,7-dione

  • MDL number MFCD31811616
  • Cas Number : 435275-07-3
  • Molecular Weight : 170.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249850&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249850.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart