AC5H3165C4DE Accela ChemBio Inc.

(5,6-Dihydro-1,4,2-dioxazin-3-yl)(o-tolyl)methanone O-Methyl Oxime

  • MDL number MFCD31811626
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (Z)-(5,6-dihydro-1,4,2-dioxazin-3-yl)(o-tolyl)methanone O-methyl oxime
Inchi Code 1S/C12H14N2O3/c1-9-5-3-4-6-10(9)11(13-15-2)12-14-17-8-7-16-12/h3-6H,7-8H2,1-2H3/b13-11-
Inchi Key PSJZACIZFMFCAQ-QBFSEMIESA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250018&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250018.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart