AC5H3165C4E0 Accela ChemBio Inc.

6-Amino-N-[2-(dimethylamino)-1-azavinyl]pyridine-2-carboxamide

  • MDL number MFCD31811628
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250221&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250221.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart