AC5H3165C4E5 Accela ChemBio Inc.

1-(2,6-Diisopropylphenyl)-3-methyl-1H-1,2,4-triazole

  • MDL number MFCD31811632
  • Cas Number : 1445975-61-0
  • Molecular Weight : 243.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,6-diisopropylphenyl)-3-methyl-1H-1,2,4-triazole

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250783&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250783.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart