AC5H49B838AC Accela ChemBio Inc.

(2-Bromo-4-methylphenyl)(2-chloro-4-fluorophenyl)methanone

  • MDL number MFCD32068400
  • Cas Number : 2226650-44-6
  • Molecular Weight : 327.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid, or colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249866&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249866.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart