AC5H49B838AE Accela ChemBio Inc.

2-(2-Methoxyphenyl)-2-[(4-tetrahydropyranyl)oxy]ethyl Methanesulfonate

  • MDL number MFCD31814470
  • Molecular Weight : 330.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250055&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250055.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart