AC5H49B838AF Accela ChemBio Inc.

Methyl 4-Benzyl-5-methyl-1H-imidazole-2-carboxylate

  • Molecular Weight : 230.26246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250532&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250532.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart