AC5H49B838B2 Accela ChemBio Inc.

Ethyl 5-Chloro-4-ethoxypyridine-2-acetate

  • Molecular Weight : 243.68676

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251380&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251380.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart