AC5H49B838B3 Accela ChemBio Inc.

1-(1-Methyl-4-imidazolyl)ethanamine Hydrochloride

  • MDL number MFCD31978034
  • Molecular Weight : 161.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251402&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251402.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart