AC5H49B838B5 Accela ChemBio Inc.

Ethyl 4-[4-(tert-Butyl)phenyl]-3,5-dimethyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD32068407
  • Molecular Weight : 299.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY252746&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY252746.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart