AC5H49B838B7 Accela ChemBio Inc.

2,2'-(1,4-Phenylene)dinicotinic Acid

  • Molecular Weight : 320.29888

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253092&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253092.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart