AC5H49B838DC Accela ChemBio Inc.

2-Amino-4,5-dichloropyrimidine

  • MDL number MFCD13193667
  • Cas Number : 403854-21-7
  • Molecular Weight : 163.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY031938&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY031938.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart