AC5H49B838E3 Accela ChemBio Inc.

trans-N,N-Dimethyl-1,2-diaminocyclohexane

  • MDL number MFCD09838678
  • Cas Number : 67198-21-4
  • Molecular Weight : 142.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY037475&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY037475.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart