AC5H49B8390A Accela ChemBio Inc.

Ethyl 3-[4-Amino-2-(methylthio)-5-thiazolyl]-3-oxopropanoate

  • MDL number MFCD03068914
  • Cas Number : 65095-75-2
  • Molecular Weight : 260.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY063929&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY063929.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart