AC5H49B83915 Accela ChemBio Inc.

6-(3,3-Dimethylallylamino)-9-(beta-D-ribofuranosyl)purine

  • MDL number MFCD00005741
  • Cas Number : 7724-76-7
  • Molecular Weight : 335.35834

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY077310&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY077310.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart