AC5H49B8391C Accela ChemBio Inc.

3-Aminopropanamide

  • MDL number MFCD00038161
  • Cas Number : 4726-85-6
  • Molecular Weight : 88.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY101590&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY101590.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart