AC5H49B83928 Accela ChemBio Inc.

4-Bromo-2-fluoro-5-nitrobenzaldehyde

  • MDL number MFCD10758067
  • Cas Number : 679839-39-5
  • Molecular Weight : 248.0060232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY110038&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY110038.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart