AC5H49B8392C Accela ChemBio Inc.

Rhodium(III) Chloride Trihydrate

  • MDL number MFCD00149840
  • Cas Number : 13569-65-8
  • Molecular Weight : 263.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY113105&attname=GoodsCode

Safety Information

hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY113105.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart