AC5H49B83931 Accela ChemBio Inc.

1-[2-(Trifluoromethyl)phenyl]cyclobutanecarboxylic Acid

  • MDL number MFCD11037069
  • Cas Number : 151157-52-7
  • Molecular Weight : 244.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY119880&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY119880.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart